Ethyl-1-methanesulfonylcyclopropane-1-carboxylate

Supplier: Matrix Scientific

067172-500MG 067172-1G
101933-386EA 403.08 USD
101933-386 101933-388
Ethyl-1-methanesulfonylcyclopropane-1-carboxylate
Ethyl-1-methanesulfonylcyclopropane-1-carboxylate

Hazard: Irritant

Formula: C₇H₁₂O₄S
MW: 192.24 g/mol
MDL Number: MFCD20921582
CAS Number: 1257236-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR