Ethyl-4-chloro-6-methylpyridine-2-carboxylate

Supplier: Matrix Scientific

067167-500MG 067167-1G
101933-366EA 600 USD
101933-366 101933-368
Ethyl-4-chloro-6-methylpyridine-2-carboxylate
Ethyl-4-chloro-6-methylpyridine-2-carboxylate

Hazard: Irritant

Formula: C₉H₁₀ClNO₂
MW: 199.64 g/mol
MDL Number: MFCD09839163
CAS Number: 315494-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR