Ethyl-6-(4-benzylpiperazin-1-yl)pyridine-2-carboxylate

Supplier: Matrix Scientific

067166-500MG 067166-1G
101933-362EA 803.08 USD
101933-362 101933-364
Ethyl-6-(4-benzylpiperazin-1-yl)pyridine-2-carboxylate
Ethyl-6-(4-benzylpiperazin-1-yl)pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₉H₂₃N₃O₂
MW: 325.41 g/mol
MDL Number: MFCD20921602
CAS Number: 1353878-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR