1-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]methyl}piperidin-4-one
Supplier: Matrix Scientific
067150-500MG
067150-1G
101933-306EA
600
USD
101933-306
101933-308
1-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]methyl}piperidin-4-one
1-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]methyl}piperidin-4-one
Hazard: Irritant
Formula:
C₁₂H₁₂ClF₃N₂O |
MDL Number:
MFCD20921567 CAS Number: 1057282-45-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...