1-(4-Chloro-6-methylpyridin-2-yl)piperazine

Supplier: Matrix Scientific

067142-500MG 067142-1G
101933-272EA 600 USD
101933-272 101933-274
1-(4-Chloro-6-methylpyridin-2-yl)piperazine
1-(4-Chloro-6-methylpyridin-2-yl)piperazine

Hazard: Irritant

Formula: C₁₀H₁₄ClN₃
MDL Number: MFCD20921601
CAS Number: 1353878-02-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR