Benzyl-4-{[(3,4-dimethoxyphenyl)amino]methyl}-4-hydroxypiperidine-1-carboxylate
Supplier: Matrix Scientific
067129-500MG
067129-1G
101933-220EA
600
USD
101933-220
101933-222
Benzyl-4-{[(3,4-dimethoxyphenyl)amino]methyl}-4-hydroxypiperidine-1-carboxylate
Benzyl-4-{[(3,4-dimethoxyphenyl)amino]methyl}-4-hydroxypiperidine-1-carboxylate
Hazard: Irritant
Formula:
C₂₂H₂₈N₂O₅ MW: 400.48 g/mol |
MDL Number:
MFCD20921586 CAS Number: 1353877-97-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...