1,2,4-Thiadiazol-5-amine hydrochloride

Supplier: Matrix Scientific

067120-1G 067120-5G
101933-198EA 29.23 USD
101933-198 101933-200
1,2,4-Thiadiazol-5-amine hydrochloride
1,2,4-Thiadiazol-5-amine hydrochloride

Hazard: Irritant

Formula: C₂H₄ClN₃S
MDL Number: MFCD19441985
CAS Number: 152513-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR