3-Isoxazolecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: Isoxazole-3-carboxylic Acid

067116-1G 067116-5G
101933-196EA 90.77 USD
101933-196 102663-828
3-Isoxazolecarboxylic acid ≥95%
3-Isoxazolecarboxylic acid

Hazard: Irritant

Formula: C₄H₃NO₃
MW: 113.07 g/mol
MDL Number: MFCD06738796
CAS Number: 3209-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR