5-Bromo-2-methylpyrimidine ≥98%
Supplier: Matrix Scientific
067101-250MG
067101-5G
101933-172EA
353.85
USD
101933-172
102551-696
5-Bromo-2-methylpyrimidine ≥98%
5-Bromo-2-methylpyrimidine
Hazard: Irritant/Store in Freezer
Formula:
C₅H₅BrN₂ MW: 173.01 g/mol |
MDL Number:
MFCD07375143 CAS Number: 7752-78-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...