3-Propionylpyridine

Supplier: Matrix Scientific

Synonyms: 1-(Pyridin-3-yl)propan-1-one

067075-1G 067075-5G
101933-142EA 60 USD
101933-142 101933-144
3-Propionylpyridine
3-Propionylpyridine

Hazard: Irritant

Formula: C₈H₉NO
MW: 135.17 g/mol
MDL Number: MFCD00023589
CAS Number: 1570-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR