Ethyl-5-chloronicotinate ≥98%
Supplier: Matrix Scientific
Synonyms:
Ethyl 3-chloro-5-pyridinecarboxylate
067069-1G
067069-5G
101933-128EA
75.38
USD
101933-128
101933-130
Ethyl-5-chloronicotinate ≥98%
Ethyl-5-chloronicotinate
Hazard: Irritant
Formula:
C₈H₈ClNO₂ MW: 185.61 g/mol |
MDL Number:
MFCD01692171 CAS Number: 20825-98-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...