Methyl-5-chloronicotinate ≥98%

Supplier: Matrix Scientific

067068-1G 067068-5G
101933-124EA 70.77 USD
101933-124 101933-126
Methyl-5-chloronicotinate ≥98%
Methyl-5-chloronicotinate

Hazard: Irritant

Formula: C₇H₆ClNO₂
MDL Number: MFCD09702467
CAS Number: 51269-81-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR