Ethane sulfinic acid sodium salt ≥90%

Supplier: Matrix Scientific

067067-1G 067067-5G
101933-120EA 129.23 USD
101933-120 101933-122
Ethane sulfinic acid sodium salt ≥90%
Ethane sulfinic acid sodium salt

Hazard: Irritant

Formula: C₂H₅NaO₂S
Melting Pt: 292 °C
Density: 1.372
MDL Number: MFCD06659476
CAS Number: 20035-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR