4-Bromo-3-chlorobenzotrifluoride ≥98%

Supplier: Matrix Scientific

Synonyms: 1-Bromo-2-chloro-4-trifluoromethylbenzene

067062-5G 067062-25G
101933-098EA 18.46 USD
101933-098 101933-100
4-Bromo-3-chlorobenzotrifluoride ≥98%
4-Bromo-3-chlorobenzotrifluoride

Hazard: Irritant-Harmful

Formula: C₇H₃BrClF₃
MW: 259.45 g/mol
MDL Number: MFCD00673985
CAS Number: 402-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR