3-Bromo-5-chlorobenzotrifluoride ≥98%

Supplier: Matrix Scientific

Synonyms: 1-Bromo-3-chloro-5-(trifluoromethyl)benzene

067061-5G 067061-25G
101933-094EA 15.38 USD
101933-094 101933-096
3-Bromo-5-chlorobenzotrifluoride ≥98%
3-Bromo-5-chlorobenzotrifluoride

Hazard: Irritant-Harmful

Formula: C₇H₃BrClF₃
MW: 259.45 g/mol
MDL Number: MFCD09260894
CAS Number: 928783-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR