2-Chloro-7H-pyrrolo[2,3-d]pyrimidine ≥95%

Supplier: Matrix Scientific

067058-1G 067058-5G
101933-086EA 330.77 USD
101933-086 101933-088
2-Chloro-7H-pyrrolo[2,3-d]pyrimidine ≥95%
2-Chloro-7H-pyrrolo[2,3-d]pyrimidine

Hazard: Irritant

Formula: C₆H₄ClN₃
MW: 153.5700 g/mol
MDL Number: MFCD07787392
CAS Number: 335654-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR