tert-Butyl-2,6-diazaspiro[3.3]heptane-2-carboxylate oxalate ≥95%
Supplier: Matrix Scientific
Synonyms:
2,6-Diaza-spiro[3.3]heptane-2-carboxylic acid tert-butyl ester oxalate salt, tert-Butyl 2,6-diazaspiro[3.3]heptane-2-carboxylate oxalate
067052-250MG
067052-1G
101933-072EA
301.54
USD
101933-072
101933-074
tert-Butyl-2,6-diazaspiro[3.3]heptane-2-carboxylate oxalate ≥95%
tert-Butyl-2,6-diazaspiro[3.3]heptane-2-carboxylate oxalate
Hazard: Irritant
Formula:
C₁₂H₂₀N₂O₆ MW: 288.3 g/mol |
MDL Number:
MFCD11976177 CAS Number: 1227382-01-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...