4-Fluoro-3-methylthiophenol ≥95%

Supplier: Matrix Scientific

067014-500MG 067014-1G 067014-5G
101932-972EA 193.85 USD
101932-972 101932-974 101932-976
4-Fluoro-3-methylthiophenol ≥95%
4-Fluoro-3-methylthiophenol

Hazard: Irritant

Formula: C₇H₇FS
MDL Number: MFCD06201757
CAS Number: 845790-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR