3-Chloro-2-(1H-pyrrol-1-yl)-5-(trifluoromethyl)pyridine ≥95%

Supplier: Matrix Scientific

067003-500MG 067003-1G 067003-5G
101932-932EA 193.85 USD
101932-932 101932-934 101932-936
3-Chloro-2-(1H-pyrrol-1-yl)-5-(trifluoromethyl)pyridine ≥95%
3-Chloro-2-(1H-pyrrol-1-yl)-5-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₁₀H₆ClF₃N₂
MDL Number: MFCD00105192
CAS Number: 287978-92-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR