2-(1-Benzylpiperidin-4-yl)acetonitrile ≥95%

Supplier: Matrix Scientific

066992-500MG 066992-1G
101932-886EA 353.85 USD
101932-886 101932-888
2-(1-Benzylpiperidin-4-yl)acetonitrile ≥95%
2-(1-Benzylpiperidin-4-yl)acetonitrile

Hazard: Irritant

Formula: C₁₄H₁₈N₂
MDL Number: MFCD00010108
CAS Number: 78056-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR