Benzyl-4-{[(2,4-dichlorophenyl)amino]methyl}-4-hydroxypiperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

066990-500MG 066990-1G
101932-880EA 600 USD
101932-880 101932-882
Benzyl-4-{[(2,4-dichlorophenyl)amino]methyl}-4-hydroxypiperidine-1-carboxylate ≥95%
Benzyl-4-{[(2,4-dichlorophenyl)amino]methyl}-4-hydroxypiperidine-1-carboxylate

Hazard: Irritant

Formula: C₂₀H₂₂Cl₂N₂O₃
MDL Number: MFCD20444427
CAS Number: 1353878-14-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR