(2S)-2-Amino-N,N-dimethyl-2-phenylacetamide hydrochloride ≥95%

Supplier: Matrix Scientific

066989-500MG 066989-1G
101932-876EA 600 USD
101932-876 101932-878
(2S)-2-Amino-N,N-dimethyl-2-phenylacetamide hydrochloride ≥95%
(2S)-2-Amino-N,N-dimethyl-2-phenylacetamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₅ClN₂O
MDL Number: MFCD20444426
CAS Number: 367259-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR