2,5-Dibromophenylacetic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 2-(2,5-Dibromophenyl)acetic acid

066946-1G 066946-5G 066946-25G
101932-828EA 36.92 USD
101932-828 101932-830 101932-832
2,5-Dibromophenylacetic acid ≥98%
2,5-Dibromophenylacetic acid

Hazard: Irritant

Formula: C₈H₆Br₂O₂
MW: 293.95 g/mol
MDL Number: MFCD02664691
CAS Number: 203314-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR