4-Bromo-3-fluorophenylacetic acid ≥98%

Supplier: Matrix Scientific

066943-1G 066943-5G
101932-824EA 29.23 USD
101932-824 101932-826
4-Bromo-3-fluorophenylacetic acid ≥98%
4-Bromo-3-fluorophenylacetic acid

Hazard: Irritant

Formula: C₈H₆BrFO₂
MW: 233.0300 g/mol
MDL Number: MFCD12756200
CAS Number: 942282-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR