Methyl-4-{[cis-2,6-dimethylmorpholin-4-yl]methyl}benzoate ≥95%

Supplier: Matrix Scientific

066927-500MG 066927-1G
101932-780EA 298.46 USD
101932-780 101932-782
Methyl-4-{[cis-2,6-dimethylmorpholin-4-yl]methyl}benzoate ≥95%
Methyl-4-{[cis-2,6-dimethylmorpholin-4-yl]methyl}benzoate

Hazard: Irritant

Formula: C₁₅H₂₁NO₃
MW: 263.34 g/mol
MDL Number: MFCD20232915
CAS Number: 1335220-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR