2-Bromo-2',5'-difluoroacetophenone ≥98%

Supplier: Matrix Scientific

Synonyms: 2,5-Difluorophenacyl bromide

066910-1G 066910-5G 066910-25G
101932-726EA 44.62 USD
101932-726 101932-728 101932-730
2-Bromo-2',5'-difluoroacetophenone ≥98%
2-Bromo-2',5'-difluoroacetophenone

Hazard: Irritant/Lachrymator

Formula: C₈H₅BrF₂O
MDL Number: MFCD00142839
CAS Number: 258513-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR