3-Bromo-4-chloropyridine hydrochloride ≥95%

Supplier: Matrix Scientific

066907-5G 066907-25G
101932-722EA 63.08 USD
101932-722 101932-724
3-Bromo-4-chloropyridine hydrochloride ≥95%
3-Bromo-4-chloropyridine hydrochloride

Hazard: Irritant

Formula: C₅H₄BrCl₂N
MDL Number: MFCD08448229
CAS Number: 181256-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR