3-Amino-4-methylbenzonitrile ≥98%

Supplier: Matrix Scientific

066906-5G 066906-25G
101932-718EA 20 USD
101932-718 101932-720
3-Amino-4-methylbenzonitrile ≥98%
3-Amino-4-methylbenzonitrile

Hazard: Irritant

Formula: C₈H₈N₂
MW: 132.1600 g/mol
MDL Number: MFCD04039781
CAS Number: 60710-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR