4-Iodo-2-(trifluoromethyl)aniline ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Amino-5-iodobenzotrifluoride

066903-1G 066903-5G 066903-25G
101932-708EA 29.23 USD
101932-708 101932-710 101932-712
4-Iodo-2-(trifluoromethyl)aniline ≥98%
4-Iodo-2-(trifluoromethyl)aniline

Hazard: Irritant

Formula: C₇H₅F₃IN
MDL Number: MFCD04972657
CAS Number: 97760-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR