4-Amino-2-fluorobenzonitrile ≥98%

Supplier: Matrix Scientific

066902-1G 066902-5G
101932-704EA 32.31 USD
101932-704 101932-706
4-Amino-2-fluorobenzonitrile ≥98%
4-Amino-2-fluorobenzonitrile

Hazard: Irritant

Formula: C₇H₅FN₂
MW: 136.1300 g/mol
MDL Number: MFCD07368709
CAS Number: 53312-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR