Benzyl-4-{[(4-fluorophenyl)amino]methyl}-4-hydroxypiperidine-1-carboxylate ≥95%
Supplier: Matrix Scientific
066868-500MG
066868-1G
101932-560EA
600
USD
101932-560
101932-562
Benzyl-4-{[(4-fluorophenyl)amino]methyl}-4-hydroxypiperidine-1-carboxylate ≥95%
Benzyl-4-{[(4-fluorophenyl)amino]methyl}-4-hydroxypiperidine-1-carboxylate
Hazard: Irritant
Formula:
C₂₀H₂₃FN₂O₃ MW: 358.42 g/mol |
MDL Number:
MFCD19982786 CAS Number: 1047655-93-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...