Benzyl-4-{[(4-chloro-2-fluorophenyl)amino]methyl}-4-hydroxypiperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

066867-500MG 066867-1G
101932-556EA 600 USD
101932-556 101932-558
Benzyl-4-{[(4-chloro-2-fluorophenyl)amino]methyl}-4-hydroxypiperidine-1-carboxylate ≥95%
Benzyl-4-{[(4-chloro-2-fluorophenyl)amino]methyl}-4-hydroxypiperidine-1-carboxylate

Hazard: Irritant

Formula: C₂₀H₂₂ClFN₂O₃
MW: 392.86 g/mol
MDL Number: MFCD19982787
CAS Number: 1330583-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR