2-Amino-3-fluorobenzonitrile ≥98%
Supplier: Matrix Scientific
066864-1G
066864-5G
101932-548EA
15.38
USD
101932-548
101932-550
2-Amino-3-fluorobenzonitrile ≥98%
2-Amino-3-fluorobenzonitrile
Hazard: Toxic
Formula:
C₇H₅FN₂ MW: 136.13 g/mol Melting Pt: 79-80°C |
MDL Number:
MFCD07368774 CAS Number: 115661-37-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...