2-(4-Methylpiperazin-1-yl)ethylaminetrihydrochloride

Supplier: Matrix Scientific

066855-1G 066855-5G
101932-528EA 47.69 USD
101932-528 101932-530
2-(4-Methylpiperazin-1-yl)ethylaminetrihydrochloride
2-(4-Methylpiperazin-1-yl)ethylaminetrihydrochloride

Hazard: Irritant

Formula: C₇H₂₀Cl₃N₃
MW: 252.62 g/mol
MDL Number: MFCD08437659
CAS Number: 90482-07-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR