Pyrrolidin-1-ylacetic acid hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(Pyrrolidin-1-yl)acetic acid hydrochloride

066852-1G 066852-5G
101932-524EA 52.31 USD
101932-524 101932-526
Pyrrolidin-1-ylacetic acid hydrochloride ≥95%
Pyrrolidin-1-ylacetic acid hydrochloride

Hazard: Irritant

Formula: C₆H₁₂ClNO₂
MW: 165.62 g/mol
MDL Number: MFCD06801274
CAS Number: 6628-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR