[4-(tert-Butylsulfanyl)phenyl]methanol ≥95%

Supplier: Matrix Scientific

066813-500MG 066813-1G 066813-5G
101932-452EA 600 USD
101932-452 101932-454 101932-456
[4-(tert-Butylsulfanyl)phenyl]methanol ≥95%
[4-(tert-Butylsulfanyl)phenyl]methanol

Hazard: Irritant

Formula: C₁₁H₁₆OS
MW: 196.31 g/mol
MDL Number: MFCD18890812
CAS Number: 500530-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR