tert-Butyl-4-(oxiran-2-ylmethyl)piperazine-1-carboxylate ≥95%

Supplier: Matrix Scientific

066812-500MG 066812-1G 066812-5G
101932-446EA 193.85 USD
101932-446 101932-448 101932-450
tert-Butyl-4-(oxiran-2-ylmethyl)piperazine-1-carboxylate ≥95%
tert-Butyl-4-(oxiran-2-ylmethyl)piperazine-1-carboxylate

Hazard: Irritant

Formula: C₁₂H₂₂N₂O₃
MDL Number: MFCD04973491
CAS Number: 159873-06-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR