(3-Fluoro-2-pyridinyl)methanol ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Fluoro-2-(hydroxymethyl)pyridine, (3-Fluoropyrid-2-yl)methanol

066802-1G 066802-5G
101932-410EA 133.85 USD
101932-410 101932-412
(3-Fluoro-2-pyridinyl)methanol ≥95%
(3-Fluoro-2-pyridinyl)methanol

Hazard: Irritant

Formula: C₆H₆FNO
MW: 127.1200 g/mol
MDL Number: MFCD09025826
CAS Number: 31181-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR