Ethyl-3-azabicyclo[3.1.0]hexane-2-carboxylate ≥95%

Supplier: Matrix Scientific

066795-500MG 066795-1G
101932-378EA 684.62 USD
101932-378 101932-380
Ethyl-3-azabicyclo[3.1.0]hexane-2-carboxylate ≥95%
Ethyl-3-azabicyclo[3.1.0]hexane-2-carboxylate

Hazard: Irritant

Formula: C₈H₁₃NO₂
MDL Number: MFCD19686125
CAS Number: 75715-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR