Benzyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate ≥95%

Supplier: Matrix Scientific

066786-500MG 066786-1G
101932-344EA 215.38 USD
101932-344 101932-346
Benzyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate ≥95%
Benzyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₇NO₃
MDL Number: MFCD11976640
CAS Number: 77211-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR