Benzyl-4-hydroxy-4-[(phenylamino)methyl]piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

066785-500MG 066785-1G 066785-5G
101932-338EA 600 USD
101932-338 101932-340 101932-342
Benzyl-4-hydroxy-4-[(phenylamino)methyl]piperidine-1-carboxylate ≥95%
Benzyl-4-hydroxy-4-[(phenylamino)methyl]piperidine-1-carboxylate

Hazard: Irritant

Formula: C₂₀H₂₄N₂O₃
MW: 340.43 g/mol
MDL Number: MFCD19686119
CAS Number: 77211-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR