1-Benzyloxy-2-nitrobenzene ≥95%

Supplier: Matrix Scientific

066638-1G 066638-5G
101932-042EA 107.69 USD
101932-042 101932-044
1-Benzyloxy-2-nitrobenzene ≥95%
1-Benzyloxy-2-nitrobenzene

Hazard: Irritant-Harmful

Formula: C₁₃H₁₁NO₃
MDL Number: MFCD00143680
CAS Number: 4560-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR