2-Methyl-5-(trifluoromethyl)benzaldehyde ≥97%

Supplier: Matrix Scientific

066632-1G 066632-5G
101932-028EA 167.69 USD
101932-028 101932-030
2-Methyl-5-(trifluoromethyl)benzaldehyde ≥97%
2-Methyl-5-(trifluoromethyl)benzaldehyde

Hazard: Irritant

Formula: C₉H₇F₃O
MW: 188.15 g/mol
MDL Number: MFCD06660200
CAS Number: 886498-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR