4-Bromo-2-(trifluoromethoxy)anisole ≥95%

Supplier: Matrix Scientific

066627-1G 066627-5G
101932-010EA 152.31 USD
101932-010 101932-012
4-Bromo-2-(trifluoromethoxy)anisole ≥95%
4-Bromo-2-(trifluoromethoxy)anisole

Hazard: Irritant

Formula: C₈H₆BrF₃O₂
MW: 271.04 g/mol
MDL Number: MFCD09907950
CAS Number: 853771-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR