5-Bromo-2-methylpyridin-3-amine ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Amino-5-bromo-2-methylpyridine
066625-1G
066625-5G
101932-006EA
242.8
USD
101932-006
102551-688
5-Bromo-2-methylpyridin-3-amine ≥97%
5-Bromo-2-methylpyridin-3-amine
Hazard: Irritant-Harmful/Light Sensitive
Formula:
C₆H₇BrN₂ MW: 187.04 g/mol |
MDL Number:
MFCD09031418 CAS Number: 914358-73-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...