5-Methylthiazole-2-carbaldehyde

Supplier: Matrix Scientific

064677-1G 064677-5G
101928-156EA 246.15 USD
101928-156 101928-158
5-Methylthiazole-2-carbaldehyde
5-Methylthiazole-2-carbaldehyde

Hazard: Irritant

Formula: C5H5NOS
MW: 127.1643 g/mol
MDL Number: MFCD08060733
CAS Number: 13838-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR