methyl cyclobutanecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl cyclobutanecarboxylate

064600-5G 064600-25G
101928-006EA 126.15 USD
101928-006 101928-008
methyl cyclobutanecarboxylate ≥95%
methyl cyclobutanecarboxylate

Hazard: Irritant

Formula: C₆H₁₀O₂
MDL Number: MFCD00095142
CAS Number: 765-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR