2-(2H-1,3-Benzodioxol-5-yl)ethan-1-ol ≥95%

Supplier: Matrix Scientific

064063-500MG 064063-1G
101926-918EA 403.08 USD
101926-918 101926-920
2-(2H-1,3-Benzodioxol-5-yl)ethan-1-ol ≥95%
2-(2H-1,3-Benzodioxol-5-yl)ethan-1-ol

Hazard: Irritant

Formula: C₉H₁₀O₃
MDL Number: MFCD00666033
CAS Number: 6006-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR