4-Amino-1-methyl-1H-pyrazole hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Methyl-1H-pyrazol-4-amine hydrochloride
064049-1G
064049-5G
101926-888EA
30.77
USD
101926-888
102663-812
4-Amino-1-methyl-1H-pyrazole hydrochloride ≥95%
4-Amino-1-methyl-1H-pyrazole hydrochloride
Hazard: Irritant
Formula:
C₄H₈ClN₃ MW: 133.5794 g/mol |
MDL Number:
MFCD07626616 CAS Number: 127107-23-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...