6-Bromoquinazolin-4-amine

Supplier: Matrix Scientific

062605-500MG 062605-2.5G
101924-200EA 380 USD
101924-200 101924-202
6-Bromoquinazolin-4-amine
6-Bromoquinazolin-4-amine

Hazard: Irritant

Formula: C₈H₆BrN₃
MW: 224.0600 g/mol
MDL Number: MFCD02228849
CAS Number: 21419-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR